Methyl-2,6-dichlorisonicotinat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 2,6-dichloropyridine-4-carboxylate, Methyl 2,6-dichloroisonicotinate, 2,6-Dichloro-4-pyridinecarboxylic acid methyl ester

M-Clarity™ Qualitätsstufe = MQ100

675962-5G
SIAL675962-5GEA 352 EUR
SIAL675962-5G
Methyl-2,6-dichlorisonicotinat, Sigma-Aldrich®
Methyl-2,6-dichlorisonicotinat
Formel: C₇H₅Cl₂NO₂
Molecular Weight: 206,03 g/mol
MDL: MFCD00044409
CAS-Nummer: 42521-09-5
EINECS: 255-868-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR