3-Phenyl-1H-pyrazol, Sigma-Aldrich®

Lieferant: Merck

676349-5G
SIAL676349-5GEA 27.4 EUR
SIAL676349-5G
3-Phenyl-1H-pyrazol, Sigma-Aldrich®
3-Phenyl-1H-pyrazole
Formel: C₉H₈N₂
MDL: MFCD00159654
CAS-Nummer: 2458-26-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR