4-(Piperidin-1-carbonyl)phenylboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(1-Piperidinylcarbonyl)benzeneboronic acid pinacol ester, Piperidin-1-yl(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanone

M-Clarity™ Qualitätsstufe = MQ100

677787-5G
SIAL677787-5GEA 125 EUR
SIAL677787-5G
4-(Piperidin-1-carbonyl)phenylboronsäurepinakolester, Sigma-Aldrich®
4-(Piperidin-1-carbonyl)phenylboronsäurepinakolester
Formel: C₁₈H₂₆BNO₃
MDL: MFCD09027083
CAS-Nummer: 938043-31-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR