N-tert-Butyl-3,5-dimethylanilin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

677981-1G
SIAL677981-1GEA 142 EUR
SIAL677981-1G
N-tert-Butyl-3,5-dimethylanilin, Sigma-Aldrich®
N-tert-Butyl-3,5-dimethylanilin
Formel: C₁₂H₁₉N
MDL: MFCD07440135
CAS-Nummer: 110993-40-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR