1,2-Dimethoxy-1,1,2,2-tetramethyldisilan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

678163-1G
SIAL678163-1GEA 143 EUR
SIAL678163-1G
1,2-Dimethoxy-1,1,2,2-tetramethyldisilan, Sigma-Aldrich®
1,2-Dimethoxy-1,1,2,2-tetramethyldisilan
Formel: C₆H₁₈Si₂O₂
Molecular Weight: 178,38 g/mol
MDL: MFCD09038446
CAS-Nummer: 10124-62-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR