5-Methyl-2-furoylchlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

678570-1G
SIAL678570-1GEA 83.1 EUR
SIAL678570-1G
5-Methyl-2-furoylchlorid, Sigma-Aldrich®
5-Methyl-2-furoylchlorid
Formel: C₆H₅ClO₂
MDL: MFCD03421416
CAS-Nummer: 14003-11-3
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR