2-Brombenzothiazol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

678600-1G
SIAL678600-1GEA 56.3 EUR
SIAL678600-1G
2-Brombenzothiazol, Sigma-Aldrich®
2-Bromobenzothiazole
Formel: C₇H₄BrNS
Molecular Weight: 214,09 g/mol
MDL: MFCD02681887
CAS-Nummer: 2516-40-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR