4-Iod-2-(trifluormethyl)benzonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

678759-1G
SIAL678759-1GEA 41.5 EUR
SIAL678759-1G
4-Iod-2-(trifluormethyl)benzonitril, Sigma-Aldrich®
4-Iod-2-(trifluormethyl)benzonitril
Formel: C₈H₃F₃IN
Molecular Weight: 297,02 g/mol
MDL: MFCD08458091
CAS-Nummer: 101066-87-9
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR