Pyrase strips
Lieferant: Merck
67886-1KT
SIAL67886-1KTEA 62.9 EUR
SIAL67886-1KT
Pyrase strips
Mikrobiologische Testsysteme Tests zur Überprüfung von Mirkoorganismen
This kit contains plastic strips with a paper zone saturated with chromogenic substrate for the detection of pyrrolidonyl peptidase and a bottle containing a developing reagent.
  • Result of the reaction is read after 2 min.
  • Detection of pyrase is a sensitive, reliable and rapid method of wide application
Pyrase Strips are used for diagnostic test for the rapid differentiation of Enterococci from the group D Streptococci and differentiation of Streptococcus pyogenes from other haemolytic Streptococci. The Strip can also be used for the identification of Gram-positive genera, catalase negative cocci, and coccobacilli.
Llimited shelf life, expiry date on the label.
Storage temperature: 2 - 8 °C
Lieferumfang: The kit contains 50 test strips and 1,0 ml reagent.