5-Fluor-3-iodtoluol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Fluor-3-iod-1-methylbenzol, 1-Fluor-3-iod-5-methylbenzol

M-Clarity™ Qualitätsstufe = MQ100

678880-1G
SIAL678880-1GEA 81.7 EUR
SIAL678880-1G
5-Fluor-3-iodtoluol, Sigma-Aldrich®
5-Fluor-3-iodtoluol
Formel: C₇H₆FI
Molecular Weight: 236,03 g/mol
MDL: MFCD07779015
CAS-Nummer: 491862-84-1
EINECS: 254-740-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR