3-Fluor-4-iodbenzonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

678899-1G
SIAL678899-1GEA 47.6 EUR
SIAL678899-1G
3-Fluor-4-iodbenzonitril, Sigma-Aldrich®
3-Fluor-4-iodbenzonitril
Formel: C₇H₃FIN
Molecular Weight: 247,01 g/mol
MDL: MFCD07782076
CAS-Nummer: 887266-99-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR