5-Fluor-2-iodbenzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

678902-1G
SIAL678902-1GEA 129 EUR
SIAL678902-1G
5-Fluor-2-iodbenzoesäure, Sigma-Aldrich®
5-Fluor-2-iodbenzoesäure
Formel: C₇H₄FIO₂
Molecular Weight: 266,01 g/mol
MDL: MFCD00837316
CAS-Nummer: 52548-63-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR