4-Fluor-2-iodbenzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

678910-1G
SIAL678910-1GEA 39.7 EUR
SIAL678910-1G
4-Fluor-2-iodbenzoesäure, Sigma-Aldrich®
4-Fluoro-2-iodobenzoic acid
Formel: C₇H₄FIO₂
Molecular Weight: 266,01 g/mol
MDL: MFCD04108278
CAS-Nummer: 56096-89-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR