4-Iod-3-(trifluormethyl)benzonitril, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
678929-1G
SIAL678929-1GEA
58.3
EUR
SIAL678929-1G
4-Iod-3-(trifluormethyl)benzonitril, Sigma-Aldrich®
4-Iod-3-(trifluormethyl)benzonitril
Formel:
C₈H₃F₃IN Molecular Weight: 297,0158 g/mol |
MDL:
MFCD08458134 CAS-Nummer: 161320-00-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...