4-Amino-2-chlorpyrimidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Chlorpyrimidin-4-ylamin

M-Clarity™ Qualitätsstufe = MQ100

679062-1G
SIAL679062-1GEA 57 EUR
SIAL679062-1G
4-Amino-2-chlorpyrimidin, Sigma-Aldrich®
4-Amino-2-chloropyrimidine
Formel: C₄H₄ClN₃
Molecular Weight: 129,55 g/mol
MDL: MFCD00194051
CAS-Nummer: 7461-50-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR