7-Fluorisatin, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

679119-5G
SIAL679119-5GEA 102 EUR
SIAL679119-5G
7-Fluorisatin, Sigma-Aldrich®
7-Fluorisatin
Formel: C₈H₄FNO₂
MDL: MFCD01569508
CAS-Nummer: 317-20-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR