7-Bromisatin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 7-Brom-1H-indol-2,3-dion, 7-Brom-2,3-dioxoindolin, 7-Bromindolin-2,3-dion, 7-Brom-2,3-indolindion

M-Clarity™ Qualitätsstufe = MQ100

679127-1G
SIAL679127-1GEA 64.2 EUR
SIAL679127-1G
7-Bromisatin, Sigma-Aldrich®
7-Bromisatin
Formel: C₈H₄BrNO₂
Molecular Weight: 226,03 g/mol
MDL: MFCD00774354
CAS-Nummer: 20780-74-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR