3-(Brommethyl)phenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Dihydroxy(3-(brommethyl)phenyl)boran, (3-(Brommethyl)phenyl)metaborsäure, 3-(Brommethyl)benzolboronsäure

M-Clarity™ Qualitätsstufe = MQ100

679445-1G 679445-5G
SIAL679445-1GEA 62.3 EUR
SIAL679445-1G SIAL679445-5G
3-(Brommethyl)phenylboronsäure, Sigma-Aldrich®
3-(Brommethyl)phenylboronsäure
Formel: C₇H₈BBrO₂
Molecular Weight: 214.85
MDL: MFCD01632207
CAS-Nummer: 51323-43-4
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR