2-(Brommethyl)phenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (2-(Brommethyl)phenyl)metaborsäure, Dihydroxy(2-(brommethyl)phenyl)boran, 2-(Brommethyl)benzolboronsäure

M-Clarity™ Qualitätsstufe = MQ100

679453-1G 679453-5G
SIAL679453-1GEA 31.6 EUR
SIAL679453-1G SIAL679453-5G
2-(Brommethyl)phenylboronsäure, Sigma-Aldrich®
2-(Brommethyl)phenylboronsäure
Formel: C₇H₈BBrO₂
MDL: MFCD01318941
CAS-Nummer: 91983-14-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR