3'-Hydroxy-4'-methoxyacetophenon, Sigma-Aldrich®

Lieferant: Merck

679704-1G 679704-5G
SIAL679704-1GEA 91.7 EUR
SIAL679704-1G SIAL679704-5G
3'-Hydroxy-4'-methoxyacetophenon, Sigma-Aldrich®
3'-Hydroxy-4'-methoxyacetophenon
Formel: C₉H₁₀O₃
MDL: MFCD00182975
CAS-Nummer: 6100-74-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR