1-Methylimidazol-4-carbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Methyl-1H-imidazole-4-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

679720-500MG
SIAL679720-500MGEA 90.6 EUR
SIAL679720-500MG
1-Methylimidazol-4-carbonsäure, Sigma-Aldrich®
1-Methylimidazol-4-carbonsäure
Formel: C₅H₆N₂O₂
Molecular Weight: 126,11 g/mol
MDL: MFCD02179560
CAS-Nummer: 41716-18-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR