5-Chlor-2-hydroxy-3-nitropyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Chloro-3-nitropyridin-2-ol

M-Clarity™ Qualitätsstufe = MQ100

679747-1G
SIAL679747-1GEA 103 EUR
SIAL679747-1G
5-Chlor-2-hydroxy-3-nitropyridin, Sigma-Aldrich®
5-Chlor-2-hydroxy-3-nitropyridin
Formel: C₅H₃ClN₂O₃
Molecular Weight: 174,54 g/mol
MDL: MFCD00114884
CAS-Nummer: 21427-61-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR