3-Chlorpicolinsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Chloropyridine-2-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ200

680052-250MG
SIAL680052-250MGEA 89.4 EUR
SIAL680052-250MG
3-Chlorpicolinsäure, Sigma-Aldrich®
3-Chlorpicolinsäure
Formel: C₆H₄ClNO₂
Molecular Weight: 157,56 g/mol
MDL: MFCD01646325
CAS-Nummer: 57266-69-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR