3-(1-Pyrrolidinylcarbonyl)phenylboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Pyrrolidin-1-yl(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanone, 3-(1-Pyrrolidinylcarbonyl)benzeneboronic acid pinacol ester

M-Clarity™ Qualitätsstufe = MQ100

680222-1G
SIAL680222-1GEA 41.1 EUR
SIAL680222-1G
3-(1-Pyrrolidinylcarbonyl)phenylboronsäurepinakolester, Sigma-Aldrich®
3-(1-Pyrrolidinylcarbonyl)phenylboronsäurepinakolester
Formel: C₁₇H₂₄BNO₃
Molecular Weight: 301.19 g/mol
MDL: MFCD09027086
CAS-Nummer: 1073353-61-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR