4-(Morpholinomethyl)phenylboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(4-Morpholinylmethyl)benzeneboronic acid pinacol ester, 4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]morpholine, 4-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)morpholine, 4,4,5,5-Tetramethyl-2-[4-(morpholinomethyl)phenyl]-1,3,2-dioxaborolane, 4-(Morpholinomethyl)phenylboronic acid Pinacol ester

M-Clarity™ Qualitätsstufe = MQ100

680230-1G
SIAL680230-1GEA 41.6 EUR
SIAL680230-1G
4-(Morpholinomethyl)phenylboronsäurepinakolester, Sigma-Aldrich®
4-(Morpholinomethyl)phenylboronsäurepinakolester
Formel: C₁₇H₂₆BNO₃
Molecular Weight: 303,20 g/mol
MDL: MFCD04974052
CAS-Nummer: 364794-79-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR