3-(N,N-Dimethylamino)phenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Dimethylaminophenylboronic acid, 3-(Dimethylamino)benzeneboronic acid

M-Clarity™ Qualitätsstufe = MQ100

680532-10G
SIAL680532-10GEA 466 EUR
SIAL680532-10G
3-(N,N-Dimethylamino)phenylboronsäure, Sigma-Aldrich®
3-(N,N-Dimethylamino)phenylboronsäure
Formel: C₈H₁₂BNO₂
Molecular Weight: 164,9974 g/mol
MDL: MFCD01318996
CAS-Nummer: 178752-79-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR