3-Hydroxy-2-iod-4-methoxybenzaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

680907-5G
SIAL680907-5GEA 124 EUR
SIAL680907-5G
3-Hydroxy-2-iod-4-methoxybenzaldehyd, Sigma-Aldrich®
3-Hydroxy-2-iodo-4-methoxybenzaldehyde
Formel: C₈H₇IO₃
Molecular Weight: 278,439 g/mol
MDL: MFCD04129042
CAS-Nummer: 138490-94-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR