3'-Chlor-4'-hydroxyacetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(3-Chlor-4-hydroxyphenyl)ethan-1-on, 1-(3-Chlor-4-hydroxyphenyl)ethanon

681032-5G
SIAL681032-5GEA 130 EUR
SIAL681032-5G
3'-Chlor-4'-hydroxyacetophenon, Sigma-Aldrich®
3'-Chlor-4'-hydroxyacetophenon
Formel: C₈H₇ClO₂
Molecular Weight: 170,6 g/mol
MDL: MFCD00663621
CAS-Nummer: 2892-29-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR