2-Brom-4-chlorbenzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

681113-5G
SIAL681113-5GEA 11.9 EUR
SIAL681113-5G
2-Brom-4-chlorbenzoesäure, Sigma-Aldrich®
2-Bromo-4-chlorobenzoic acid
Formel: C₇H₄BrClO₂
Molecular Weight: 235,46 g/mol
MDL: MFCD00672930
CAS-Nummer: 936-08-3
UN: 2928
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR