3-(3-Bromphenyl)propionsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

681148-5G
SIAL681148-5GEA 110 EUR
SIAL681148-5G
3-(3-Bromphenyl)propionsäure, Sigma-Aldrich®
3-(3-Bromphenyl)propionsäure
Formel: C₉H₉BrO₂
Molecular Weight: 229,07 g/mol
MDL: MFCD01310792
CAS-Nummer: 42287-90-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR