2-Iodbenzonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: o-Iodobenzonitrile

M-Clarity™ Qualitätsstufe = MQ100

681172-5G
SIAL681172-5GEA 90 EUR
SIAL681172-5G
2-Iodbenzonitril, Sigma-Aldrich®
2-Iodbenzonitril
Formel: C₇H₄IN
Molecular Weight: 229,02 g/mol
MDL: MFCD00079761
CAS-Nummer: 4387-36-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR