2-(Trifluormethyl)nicotinsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Trifluoromethylnicotinic acid

M-Clarity™ Qualitätsstufe = MQ100

681261-500MG
SIAL681261-500MGEA 159 EUR
SIAL681261-500MG
2-(Trifluormethyl)nicotinsäure, Sigma-Aldrich®
2-(Trifluormethyl)nicotinsäure
Formel: C₇H₄F₃NO₂
Molecular Weight: 191,11 g/mol
MDL: MFCD01862052
CAS-Nummer: 131747-43-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR