2-Chlor-5-methoxypyrimidin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

681288-500MG
SIAL681288-500MGEA 154 EUR
SIAL681288-500MG
2-Chlor-5-methoxypyrimidin, Sigma-Aldrich®
2-Chloro-5-methoxypyrimidine
Formel: C₅H₅ClN₂O
Molecular Weight: 144,56 g/mol
MDL: MFCD08702770
CAS-Nummer: 22536-65-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR