3-(Benzyloxy)benzoesäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Benzyloxybenzoic acid

M-Clarity™ Qualitätsstufe = MQ100

681717-5G
SIAL681717-5GEA 129 EUR
SIAL681717-5G
3-(Benzyloxy)benzoesäure, Sigma-Aldrich®
3-(Benzyloxy)benzoesäure
Formel: C₁₄H₁₂O₃
Molecular Weight: 228.25 g/mol
MDL: MFCD00527901
CAS-Nummer: 69026-14-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR