6-Brom-1,3-dichlorisochinolin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

681768-1G
SIAL681768-1GEA 115 EUR
SIAL681768-1G
6-Brom-1,3-dichlorisochinolin, Sigma-Aldrich®
6-Bromo-1,3-dichloroisoquinoline
Formel: C₉H₄BrCl₂N
Molecular Weight: 276,9448 g/mol
MDL: MFCD09258639
CAS-Nummer: 552331-05-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR