1,2-Bis(2-methoxyphenyl)-1,1,2,2-tetramethyldisilan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Bis(2-methoxyphenyl)-1,1,2,2-tetramethyldisilane

M-Clarity™ Qualitätsstufe = MQ100

682233-1G
SIAL682233-1GEA 30.4 EUR
SIAL682233-1G
1,2-Bis(2-methoxyphenyl)-1,1,2,2-tetramethyldisilan, Sigma-Aldrich®
1,2-Bis(2-methoxyphenyl)-1,1,2,2-tetramethyldisilan
Formel: C₁₈H₂₆O₂Si₂
Molecular Weight: 330,57 g/mol
MDL: MFCD09265162
CAS-Nummer: 332343-84-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR