2-Nitro-6-(trifluormethyl)anilin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Amino-3-nitrobenzotrifluoride

M-Clarity™ Qualitätsstufe = MQ200

682438-1G
SIAL682438-1GEA 204 EUR
SIAL682438-1G
2-Nitro-6-(trifluormethyl)anilin, Sigma-Aldrich®
2-Nitro-6-(trifluormethyl)anilin
Formel: C₇H₅F₃N₂O₂
Molecular Weight: 206,1220 g/mol
MDL: MFCD08532494
CAS-Nummer: 24821-17-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR