4-Cyan-3-fluorbenzaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Fluoro-4-formylbenzonitrile, 4-Cyano-3-fluorobenzaldehyde

M-Clarity™ Qualitätsstufe = MQ100

682454-500MG
SIAL682454-500MGEA 124 EUR
SIAL682454-500MG
4-Cyan-3-fluorbenzaldehyd, Sigma-Aldrich®
4-Cyano-3-fluorobenzaldehyde
Formel: C₈H₄FNO
Molecular Weight: 149,1200 g/mol
MDL: MFCD06738989
CAS-Nummer: 101048-76-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR