2-Fluor-6-iodbenzaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Fluoro-2-Iodobenzaldehyde

M-Clarity™ Qualitätsstufe = MQ100

682489-1G
SIAL682489-1GEA 55 EUR
SIAL682489-1G
2-Fluor-6-iodbenzaldehyd, Sigma-Aldrich®
2-Fluoro-6-iodobenzaldehyde
Formel: C₇H₄FIO
Molecular Weight: 250,01 g/mol
MDL: MFCD07371506
CAS-Nummer: 146137-72-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR