2-Amino-4-(2,4-difluorphenyl)thiazol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

683280-1G
SIAL683280-1GEA 81 EUR
SIAL683280-1G
2-Amino-4-(2,4-difluorphenyl)thiazol, Sigma-Aldrich®
2-Amino-4-(2,4-difluorophenyl)thiazole
Formel: C₉H₆F₂N₂S
Molecular Weight: 212,22 g/mol
MDL: MFCD00472891
CAS-Nummer: 105512-80-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR