2-Amino-4-fluorphenol, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 5-Fluor-2-hydroxyanilin

M-Clarity™ Qualitätsstufe = MQ100

683302-1G
SIAL683302-1GEA 32.4 EUR
SIAL683302-1G
2-Amino-4-fluorphenol, Sigma-Aldrich®
2-Amino-4-fluorphenol
Formel: C₆H₆FNO
Molecular Weight: 127,12 g/mol
MDL: MFCD00077451
CAS-Nummer: 399-97-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR