3,5-Dimethyl-4-iod-1H-pyrazol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3,5-Dimethyl-4-iodopyrazole
M-Clarity™ Qualitätsstufe = MQ100
683523-5G
SIAL683523-5GEA
94.2
EUR
SIAL683523-5G
3,5-Dimethyl-4-iod-1H-pyrazol, Sigma-Aldrich®
3,5-Dimethyl-4-iodo-1H-pyrazole
Formel:
C₅H₇IN₂ Molecular Weight: 222.03 g/mol |
MDL:
MFCD00040247 CAS-Nummer: 2033-45-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...