1-Chlor-4-ethylbenzol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Chloro(ethylbenzene)

683744-5G
SIAL683744-5GEA 122 EUR
SIAL683744-5G
1-Chlor-4-ethylbenzol, Sigma-Aldrich®
1-Chlor-4-ethylbenzol
Formel: C₈H₉Cl
Molecular Weight: 140,61 g/mol
MDL: MFCD00040855
CAS-Nummer: 622-98-0
EINECS: 210-763-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR