1-Boc-3,5-dimethyl-1H-pyrazol-4-boronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(1-Boc-3,5-dimethyl-4-pyrazolyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylic acid tert-Butyl ester, 1-Boc-3,5-dimethylpyrazole-4-boronic acid Pinacol ester, tert-Butyl 3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylate, 1-(tert-Butoxycarbonyl)-3,5-dimethylpyrazole-4-boronic acid Pinacol ester, 2-(1-tert-Butoxycarbonyl-3,5-dimethyl-4-pyrazolyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, tert-Butyl 3,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylate, 1-(tert-Butoxycarbonyl)-3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, 1-Boc-3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole

M-Clarity™ Qualitätsstufe = MQ100

683779-1G
SIAL683779-1GEA 120 EUR
SIAL683779-1G
1-Boc-3,5-dimethyl-1H-pyrazol-4-boronsäurepinakolester, Sigma-Aldrich®
1-Boc-3,5-dimethyl-1H-pyrazol-4-boronsäurepinakolester
Formel: C₁₆H₂₇BN₂O₄
Molecular Weight: 322.2076 g/mol
MDL: MFCD09027070
CAS-Nummer: 1073354-70-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR