2-(Methoxycarbonyl)phenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Methoxycarbonylphenylboronic acid, 2-(Methoxycarbonyl)benzeneboronic acid

M-Clarity™ Qualitätsstufe = MQ100

683809-5G
SIAL683809-5GEA 90.6 EUR
SIAL683809-5G
2-(Methoxycarbonyl)phenylboronsäure, Sigma-Aldrich®
2-(Methoxycarbonyl)phenylboronsäure
Formel: C₈H₉BO₄
Molecular Weight: 179.97 g/mol
MDL: MFCD02179452
CAS-Nummer: 374538-03-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR