2-(Ethoxycarbonyl)phenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(Ethoxycarbonyl)benzeneboronic acid, (2-(Ethoxycarbonyl)phenyl)boronic acid

M-Clarity™ Qualitätsstufe = MQ100

683833-5G
SIAL683833-5GEA 92.8 EUR
SIAL683833-5G
2-(Ethoxycarbonyl)phenylboronsäure, Sigma-Aldrich®
2-(Ethoxycarbonyl)phenylboronsäure
Formel: C₉H₁₁BO₄
Molecular Weight: 193.99 g/mol
MDL: MFCD02179453
CAS-Nummer: 380430-53-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR