8-Chlorchinaldin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 8-Chloro-2-methylquinoline

M-Clarity™ Qualitätsstufe = MQ100

683914-5G
SIAL683914-5GEA 86.7 EUR
SIAL683914-5G
8-Chlorchinaldin, Sigma-Aldrich®
8-Chlorchinaldin
Formel: C₁₀H₈ClN
Molecular Weight: 177.63 g/mol
MDL: MFCD00051764
CAS-Nummer: 3033-82-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR