2,4-Bis(trifluormethyl)phenylboronsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

683965-1G
SIAL683965-1GEA 37.2 EUR
SIAL683965-1G
2,4-Bis(trifluormethyl)phenylboronsäure, Sigma-Aldrich®
2,4-Bis(trifluormethyl)phenylboronsäure
Formel: C₈H₅BF₆O₂
Molecular Weight: 257,93 g/mol
MDL: MFCD01631349
CAS-Nummer: 153254-09-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR