[6,6]-Phenyl-C61-buttersäure-methylester ≥99,9%, Sigma-Aldrich®

Lieferant: Merck

Synonyme: [60]PCBM, PCBM, [6,6]-Phenyl-C61-butyric Acid Methyl Ester, 3′H-Cyclopropa[1,9] [5,6]fullerene-C60-Ih-3′-butanoic acid 3′-phenyl methyl ester, Methyl [6,6]-Phenyl-C61-butyrate

M-Clarity™ Qualitätsstufe = MQ100

684457-100MG
SIAL684457-100MGEA 945 EUR
SIAL684457-100MG
[6,6]-Phenyl-C61-buttersäure-methylester ≥99,9%, Sigma-Aldrich®
[6,6]-Phenyl-C61-butyric acid methyl ester

Soluble n-channel organic semiconductor. For use as an n-type layer in plastic electronics, especially bulk heterojunction OFETs and photovoltaic cells (PVs)


Synonyms: 1-[3-(Methoxycarbonyl)propyl]-1-phenyl-[6.6]C61, 3′H-Cyclopropa[1,9] [5,6]fullerene-C60-Ih-3′-butanoic acid 3′-phenyl methyl ester.

Formel: C₇₂H₁₄O₂
Molecular Weight: 910,90 g/mol
MDL: MFCD07784544
CAS-Nummer: 160848-22-6

Bestellen

Spezifikation Testergebnisse

Assay >99,9%
Solubility Soluble in chlorobenzene, toluene and other organic solvents
Orbital energy (HOMO) 6,1 eV
Orbital energy (LUMO) 3,7 eV

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR