6-Amino-3-cyan-2-methylpyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Amino-2-methylnicotinonitrile, 6-Amino-3-cyano-2-methylpyridine, 6-Amino-2-methylpyridine-3-carbonitrile

M-Clarity™ Qualitätsstufe = MQ100

684627-1G
SIAL684627-1GEA 84.9 EUR
SIAL684627-1G
6-Amino-3-cyan-2-methylpyridin, Sigma-Aldrich®
6-Amino-3-cyan-2-methylpyridin
Formel: C₇H₇N₃
Molecular Weight: 133,15 g/mol
MDL: MFCD05865116
CAS-Nummer: 183428-90-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR